Gene Transcripts
- Gene Symbol
- RBBP4
- Entrez Gene ID
- 5928
- Gene Name
- RB binding protein 4, chromatin remodeling factor
- Gene Aliases
- NURF55, RBAP48, lin-53
- Chromosome Location
- Chr. 1: 32651148 - 32686211 on Build GRCh38
- UniGene ID
- Hs.16003
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135255.1 |
NP_001128727.1
|
3, 4 |
471 |
|
NM_001135256.1 |
NP_001128728.1
|
3, 4 |
374 |
|
NM_005610.2 |
NP_005601.1
|
3, 4 |
474 |
|
GenBank mRNA |
AK222779.1 |
- |
3, 4 |
377 |
|
AK299251.1 |
- |
3, 4 |
374 |
|
AK312571.1 |
- |
3, 4 |
412 |
|
BC003092.1 |
- |
3, 4 |
401 |
3356558 |
BC015123.1 |
- |
1, 2 |
135 |
|
BC053904.1 |
- |
3, 4 |
387 |
6526200 |
BC075836.1 |
- |
3, 4 |
342 |
6338602 |
BT007309.1 |
- |
3, 4 |
315 |
|
DC315298.1 |
- |
|
475 |
|
X71810.1 |
- |
3, 4 |
388 |
|
X74262.1 |
- |
3, 4 |
399 |
|
View Full Product Details