Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025590.4 | NP_079866.2 | 16 | 2870 | |
GenBank mRNA | AK031588.1 | - | 16 | 2787 | |
AK038546.1 | - | 16 | 2809 | ||
AK048670.1 | - | 16 | 2870 | ||
BC042492.1 | - | 16 | 2805 | 4240966 |