Gene Transcripts

Gene Symbol
Crebl2
Entrez Gene ID
232430
Gene Name
cAMP responsive element binding protein-like 2
Gene Aliases
AI046348, B230205M03
Chromosome Location
Chr. 6: 134828686 - 134858931 on Build GRCm38
UniGene ID
Mm.267010
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177687.3 NP_808355.1 5 2015
GenBank mRNA AK045467.1 - 5 2016

View Full Product Details