Gene Transcripts

Gene Symbol
Ano5
Entrez Gene ID
233246
Gene Name
anoctamin 5
Gene Aliases
9330162L24, Gdd1, Tmem16e
Chromosome Location
Chr. 7: 51510875 - 51600693 on Build GRCm38
UniGene ID
Mm.254431
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271879.1 NP_001258808.1 10 1245
NM_177694.6 NP_808362.2 11 1287
GenBank mRNA AB125740.1 - 11 1287
AB206762.1 - 11 1287
AK034197.1 - 11 1269
BC109163.2 - 10 1167 40047987

View Full Product Details