Gene Transcripts

Gene Symbol
Skint4
Entrez Gene ID
320640
Gene Name
selection and upkeep of intraepithelial T cells 4
Gene Aliases
9530098N22Rik
Chromosome Location
Chr. 4: 112072016 - 112168076 on Build GRCm38
UniGene ID
Mm.253775
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178786.4 NP_848901.2 11 2767
GenBank mRNA AK028550.1 - 11 2819
AK028795.1 - 11 2768
AK162813.1 - 12 2782

View Full Product Details