Gene Transcripts

Gene Symbol
Homez
Entrez Gene ID
239099
Gene Name
homeodomain leucine zipper-encoding gene
Gene Aliases
mKIAA1443
Chromosome Location
Chr. 14: 54852736 - 54871352 on Build GRCm38
UniGene ID
Mm.280994
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177705.1 NP_001171176.1 2 428
NM_183174.3 NP_898997.2 2 273
GenBank mRNA AK129361.1 - 2 318
AY258064.1 - 2 376
BC094669.1 - 2 273 30616315
CJ068402.1 - 2 389

View Full Product Details