Gene Transcripts

Gene Symbol
Rnf26
Entrez Gene ID
213211
Gene Name
ring finger protein 26
Gene Aliases
8030450I18Rik
Chromosome Location
Chr. 9: 44110781 - 44113051 on Build GRCm38
UniGene ID
Mm.392032
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153762.3 NP_717095.2 1 926
GenBank mRNA AK033164.1 - 1 1380
AK085078.1 - 1376
AK157821.1 - 1 1375
BC004739.1 - 1 977 3501511
BC063251.1 - 1 926 6413980
BC111036.1 - 2 915

View Full Product Details