Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001291047.1 | NP_001277976.1 | 5 | 773 | |
NM_001291053.1 | NP_001277982.1 | 3 | 367 | ||
NM_023249.6 | NP_075738.1 | 4 | 647 | ||
GenBank mRNA | AB098740.1 | - | 3 | 259 | |
AF190624.1 | - | 4 | 563 | ||
AK005955.1 | - | 4 | 483 | ||
AK006049.1 | - | 4 | 483 | ||
AK006137.1 | - | 3 | 265 | ||
AK006505.1 | - | 4 | 458 | ||
AK006858.1 | - | 4 | 535 | ||
AK011452.1 | - | 4 | 578 | ||
AK014931.1 | - | 4 | 473 | ||
AK015752.1 | - | 3 | 484 | ||
AK018910.1 | - | 4 | 458 | ||
AK019819.1 | - | 4 | 478 | ||
AK076063.1 | - | 4 | 590 | ||
AK133401.1 | - | 4 | 561 | ||
AK139453.1 | - | 4 | 555 | ||
BB610926.1 | - | 5 | 686 | ||
BC049737.1 | - | 3 | 277 | 6771362 | |
BQ573264.1 | - | 3 | 333 |