Gene Transcripts
- Gene Symbol
- PRCC
- Entrez Gene ID
- 5546
- Gene Name
- papillary renal cell carcinoma (translocation-associated)
- Gene Aliases
- RCCP1, TPRC
- Chromosome Location
- Chr. 1: 156767482 - 156800819 on Build GRCh38
- UniGene ID
- Hs.516948
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005973.4 |
NP_005964.3
|
1, 2 |
760 |
|
XM_005245313.2 |
XP_005245370.1
|
1, 2 |
760 |
|
XM_005245314.1 |
XP_005245371.1
|
1, 2 |
760 |
|
XM_011509726.1 |
XP_011508028.1
|
1, 2 |
760 |
|
GenBank mRNA |
AK289872.1 |
- |
1, 2 |
700 |
|
BC004913.2 |
- |
1, 2 |
676 |
3535413 |
BC010450.1 |
- |
1, 2 |
688 |
4179105 |
BM423590.1 |
- |
|
746 |
|
KU178273.1 |
- |
1, 2 |
470 |
|
KU178274.1 |
- |
1, 2 |
470 |
|
KU178275.1 |
- |
1, 2 |
470 |
|
X97124.1 |
- |
1, 2 |
660 |
|
View Full Product Details