Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133217.3 | NP_573480.1 | 3, 4 | 527 | |
GenBank mRNA | AJ290392.1 | - | 2, 3 | 370 | |
BB624957.1 | - | 3, 4 | 572 | ||
BC107006.2 | - | 2, 3 | 370 | 40046913 | |
BC107007.2 | - | 2, 3 | 370 | 40046914 |