Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005898.4 | NP_005889.3 | 4 | 497 | |
NM_203364.2 | NP_976240.1 | 4 | 497 | ||
XM_017017749.1 | XP_016873238.1 | 3 | 468 | ||
GenBank mRNA | AK127972.1 | - | 3 | 297 | |
AK299720.1 | - | 3 | 372 | ||
BC001731.1 | - | 4 | 497 | 3355481 | |
Z48042.1 | - | 3 | 350 |