Gene Transcripts

Gene Symbol
Brms1
Entrez Gene ID
107392
Gene Name
breast cancer metastasis-suppressor 1
Gene Aliases
AV003220, AW554636
Chromosome Location
Chr. 19: 5041236 - 5049917 on Build GRCm38
UniGene ID
Mm.29628
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134155.1 NP_598916.1 7 648
GenBank mRNA AF233580.1 - 7 659
AK076729.1 - 7 656
BC016108.1 - 7 648 4921658
BC038380.1 - 2887

View Full Product Details