Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_134437.3 | NP_602319.1 | 5 | 629 | |
GenBank mRNA | AF424804.1 | - | 1 | 58 | |
AF459444.1 | - | 5 | 548 | ||
AF494209.1 | - | 1 | 338 | ||
AF494210.1 | - | 5 | 689 | ||
AK133233.1 | - | 7 | 792 | ||
AK139904.1 | - | 5 | 554 | ||
BC066804.1 | - | 4 | 1039 | ||
BC138629.1 | - | 5 | 584 | 8861649 |