Gene Transcripts

Gene Symbol
Slain1
Entrez Gene ID
105439
Gene Name
SLAIN motif family, member 1
Gene Aliases
9630044O09Rik, AA675320, AW742596
Chromosome Location
Chr. 14: 103650071 - 103704912 on Build GRCm38
UniGene ID
Mm.27548
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198014.2 NP_932131.2 6 1640
GenBank mRNA AK036190.1 - 8 1745
BC025223.1 - 4 794 5010865
BC079866.1 - 6 1640 6811096

View Full Product Details