Gene Transcripts

Gene Symbol
Cep78
Entrez Gene ID
208518
Gene Name
centrosomal protein 78
Gene Aliases
5730599I05Rik, D030027P05
Chromosome Location
Chr. 19: 15955768 - 15985016 on Build GRCm38
UniGene ID
Mm.105652
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198019.2 NP_932136.2 2 472
GenBank mRNA AK050870.1 - 2 179
BC070400.1 - 2 472 5694341
BC071270.1 - 2 456

View Full Product Details