Gene Transcripts

Gene Symbol
Calm4
Entrez Gene ID
80796
Gene Name
calmodulin 4
Gene Aliases
2310037J09Rik, AI649108, DD112, Scarf
Chromosome Location
Chr. 13: 3837757 - 3838671 on Build GRCm38
UniGene ID
Mm.21075
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020036.4 NP_064420.2 1 885
GenBank mRNA AK003648.1 - 1 886
AK009664.1 - 1 887
AK009956.1 - 1 886
AY293058.1 - 1 868
BC060284.1 - 1 871 30293362

View Full Product Details