Gene Transcripts

Gene Symbol
PCP4
Entrez Gene ID
5121
Gene Name
Purkinje cell protein 4
Gene Aliases
PEP-19
Chromosome Location
Chr. 21: 39867421 - 39929397 on Build GRCh38
UniGene ID
Hs.80296
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006198.2 NP_006189.2 3 448
GenBank mRNA AK289964.1 - 3 431
BC013791.1 - 3 424 4345740
U52969.1 - 3 448
X93349.1 - 3 425

View Full Product Details