Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023892.2 | NP_076381.1 | 2 | 616 | |
GenBank mRNA | AF296282.1 | - | 2 | 535 | |
AF296283.1 | - | 2 | 535 | ||
AK019013.1 | - | 2 | 318 | ||
BC116909.1 | - | 2 | 616 | 40126228 | |
BC116913.1 | - | 2 | 616 | 40126232 |