Gene Transcripts

Gene Symbol
Spint4
Entrez Gene ID
78239
Gene Name
serine protease inhibitor, Kunitz type 4
Gene Aliases
9230105I15Rik
Chromosome Location
Chr. 2: 164698501 - 164702448 on Build GRCm38
UniGene ID
Mm.160060
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030058.2 NP_084334.1 1, 2 172
GenBank mRNA AK020315.1 - 1, 2 145
AY226988.1 - 1, 2 172
BC119299.1 - 1, 2 142 8734048
BC119301.1 - 1, 2 142 8734050

View Full Product Details