Gene Transcripts

Gene Symbol
DNAJB1
Entrez Gene ID
3337
Gene Name
DnaJ heat shock protein family (Hsp40) member B1
Gene Aliases
HSPF1, Hdj1, Hsp40, RSPH16B, Sis1
Chromosome Location
Chr. 19: 14514764 - 14529821 on Build GRCh38
UniGene ID
Hs.515210
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300914.1 NP_001287843.1 2 435
NM_001313964.1 NP_001300893.1 3 361
NM_006145.2 NP_006136.1 2 584
XM_006722733.1 XP_006722796.1 3 374
XM_006722734.3 XP_006722797.1 3 798
XM_011527956.2 XP_011526258.1 3 353
GenBank mRNA AK301817.1 - 3 276
AK312624.1 - 2 445
BC002352.2 - 2 408 2821008
BC019827.1 - 2 398 4152608
CR536485.1 - 2 374
CR541677.1 - 2 374
D49547.1 - 2 414
DA377896.1 - 2 435
X62421.1 - 2 418

View Full Product Details