Gene Transcripts

Gene Symbol
Sacm1l
Entrez Gene ID
83493
Gene Name
SAC1 (suppressor of actin mutations 1, homolog)-like (S. cerevisiae)
Gene Aliases
SAC1, Sac1p, mKIAA0851
Chromosome Location
Chr. 9: 123529882 - 123592598 on Build GRCm38
UniGene ID
Mm.273671
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030692.4 NP_109617.1 5 448
GenBank mRNA AJ245720.1 - 5 578
AJ297526.1 - 5 449
AK089174.1 - 5 532
AK122390.1 - 5 513
AK154759.1 - 5 548
AK161111.1 - 5 482
AK171080.1 - 5 532
AK198578.1 - 2 178
BC117752.1 - 5 449 40090556
BC117753.1 - 5 449 40090558

View Full Product Details