Gene Transcripts

Gene Symbol
Tmprss5
Entrez Gene ID
80893
Gene Name
transmembrane protease, serine 5 (spinesin)
Gene Aliases
spinesin
Chromosome Location
Chr. 9: 49080439 - 49117591 on Build GRCm38
UniGene ID
Mm.72799
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB016229.1 - 10 1087
AB016230.1 - 10 1089
AB016423.1 - 6 704
AB041037.1 - 10 1046

View Full Product Details