Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_053142.3 | NP_444372.2 | 1 | 629 | |
GenBank mRNA | AF326310.1 | - | 1 | 491 | |
AK077255.1 | - | 1 | 600 | ||
AK122533.1 | - | 855 | |||
AY013778.1 | - | 1 | 491 | ||
BC017149.1 | - | 1 | 639 | 4506788 |