Gene Transcripts

Gene Symbol
Ssx2ip
Entrez Gene ID
99167
Gene Name
synovial sarcoma, X breakpoint 2 interacting protein
Gene Aliases
AU014939, AU042321, Adip
Chromosome Location
Chr. 3: 146404617 - 146440144 on Build GRCm38
UniGene ID
Mm.200783
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253768.1 NP_001240697.1 7 1153
NM_001253769.1 NP_001240698.1 6 1049
NM_001253770.1 NP_001240699.1 6 898
NM_138744.3 NP_620083.1 6 898
GenBank mRNA AF532969.1 - 5 693
AK031356.1 - 7 877
AK043865.1 - 6 1002
AK049080.1 - 6 1050
AK077112.1 - 7 938
AK173064.1 - 6 885
BC021749.1 - 6 853 4165430

View Full Product Details