Gene Transcripts
- Gene Symbol
- Dcun1d4
- Entrez Gene ID
- 100737
- Gene Name
- DCN1, defective in cullin neddylation 1, domain containing 4 (S. cerevisiae)
- Gene Aliases
- AI836376
- Chromosome Location
- Chr. 5: 73481027 - 73560794 on Build GRCm38
- UniGene ID
- Mm.220312
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001190733.1 |
NP_001177662.1
|
4, 5 |
400 |
|
NM_001190734.1 |
NP_001177663.1
|
5, 6 |
335 |
|
NM_178896.5 |
NP_849227.1
|
4, 5 |
400 |
|
GenBank mRNA |
AK033559.1 |
- |
4, 5 |
345 |
|
AK076991.1 |
- |
5, 6 |
336 |
|
BC060213.1 |
- |
4, 5 |
342 |
|
BC108346.1 |
- |
4, 5 |
302 |
6310392 |
BC137627.1 |
- |
4, 5 |
344 |
8860646 |
BC137628.1 |
- |
4, 5 |
344 |
8860647 |
BC144794.1 |
- |
4, 5 |
344 |
9053342 |
View Full Product Details