Gene Transcripts

Gene Symbol
Wdr31
Entrez Gene ID
71354
Gene Name
WD repeat domain 31
Gene Aliases
5430402I10Rik, spWD
Chromosome Location
Chr. 4: 62448653 - 62470964 on Build GRCm38
UniGene ID
Mm.155573
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290521.1 NP_001277450.1 5, 6 625
NM_001290522.1 NP_001277451.1 5, 6 515
NM_023597.2 NP_076086.1 5, 6 518
GenBank mRNA AF274321.1 - 5, 6 495
AK017259.1 - 5, 6 508
AK141026.1 - 5, 6 623
BC026500.1 - 4, 5 404
BC037102.1 - 5, 6 508 5364307
CK618879.1 - 5 625

View Full Product Details