Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027460.2 | NP_081736.2 | 7 | 1143 | |
GenBank mRNA | AK017628.1 | - | 7 | 1054 | |
AK157423.1 | - | 7 | 1144 | ||
AK181507.1 | - | 1 | 43 | ||
AK207629.1 | - | 1 | 43 | ||
AW989568.1 | - | 3 | 484 | ||
BB772382.1 | - | 2 | 240 | ||
BC011293.1 | - | 7 | 1118 | 3989313 |