Gene Transcripts

Gene Symbol
Smg9
Entrez Gene ID
71997
Gene Name
smg-9 homolog, nonsense mediated mRNA decay factor (C. elegans)
Gene Aliases
1500002O20Rik, AI429693, N28092
Chromosome Location
Chr. 7: 24398910 - 24422777 on Build GRCm38
UniGene ID
Mm.150977
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028047.2 NP_082323.1 11 1427
GenBank mRNA AK005126.1 - 11 1427
AK039368.1 - 11 1426
AK141576.1 - 11 1418
AK169651.1 - 11 1351
BC034211.1 - 11 1342 4988361

View Full Product Details