Gene Transcripts
- Gene Symbol
- SRSF7
- Entrez Gene ID
- 6432
- Gene Name
- serine and arginine rich splicing factor 7
- Gene Aliases
- 9G8, AAG3, SFRS7
- Chromosome Location
- Chr. 2: 38743599 - 38751494 on Build GRCh38
- UniGene ID
- Hs.309090
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001031684.2 |
NP_001026854.1
|
2, 3 |
442 |
|
NM_001195446.1 |
NP_001182375.1
|
2, 3 |
442 |
|
XM_005264484.2 |
XP_005264541.1
|
2, 3 |
309 |
|
XM_005264485.2 |
XP_005264542.1
|
2, 3 |
309 |
|
GenBank mRNA |
AB445102.1 |
- |
2, 3 |
204 |
|
AK091425.1 |
- |
2, 3 |
257 |
|
AK225141.1 |
- |
2, 3 |
309 |
|
AK293667.1 |
- |
2, 3 |
309 |
|
AK297161.1 |
- |
2, 3 |
309 |
|
AY166860.1 |
- |
2, 3 |
330 |
|
AY513287.1 |
- |
2, 3 |
285 |
|
BC000997.2 |
- |
2, 3 |
289 |
2901284 |
BC017369.2 |
- |
2, 3 |
300 |
2967417 |
BC017908.1 |
- |
2, 3 |
307 |
4276642 |
BC022328.1 |
- |
2, 3 |
307 |
4251314 |
BT006745.1 |
- |
2, 3 |
204 |
|
DC358626.1 |
- |
2, 3 |
443 |
|
L22253.1 |
- |
2, 3 |
309 |
|
View Full Product Details