Gene Transcripts

Gene Symbol
Mfsd8
Entrez Gene ID
72175
Gene Name
major facilitator superfamily domain containing 8
Gene Aliases
2810423E13Rik, AI836898, AV142426, Cln7
Chromosome Location
Chr. 3: 40818112 - 40846856 on Build GRCm38
UniGene ID
Mm.130610
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028140.4 NP_082416.2 12 2586
GenBank mRNA AK035291.1 - 12 2550
AK077957.1 - 12 2587
BC010483.1 - 12 2435

View Full Product Details