Gene Transcripts

Gene Symbol
Pih1d2
Entrez Gene ID
72614
Gene Name
PIH1 domain containing 2
Gene Aliases
2700059L22Rik
Chromosome Location
Chr. 9: 50617321 - 50625000 on Build GRCm38
UniGene ID
Mm.147232
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028300.2 NP_082576.1 4 680
GenBank mRNA AK012458.1 - 4 656
BC019481.1 - 4 578 4190604
BC039645.1 - 4 670 5373181

View Full Product Details