Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006387.5 | NP_006378.3 | 13 | 2264 | |
GenBank mRNA | AB209730.1 | - | 4 | 641 | |
AF536542.1 | - | 13 | 2231 | ||
AK298408.1 | - | 5 | 992 | ||
BC021294.2 | - | 13 | 2249 | 4053575 | |
U94836.2 | - | 12 | 2166 |