Gene Transcripts
- Gene Symbol
- Nsun6
- Entrez Gene ID
- 74455
- Gene Name
- NOL1/NOP2/Sun domain family member 6
- Gene Aliases
- 4933403D21Rik, 4933414E04Rik, NOPD1
- Chromosome Location
- Chr. 2: 14991522 - 15054872 on Build GRCm38
- UniGene ID
- Mm.478923
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001165941.1 |
NP_001159413.1
|
3, 4 |
668 |
|
NM_001165942.1 |
NP_001159414.1
|
2, 3 |
555 |
|
NM_001165943.1 |
NP_001159415.1
|
2, 3 |
372 |
|
NM_028950.4 |
NP_083226.3
|
2, 3 |
555 |
|
GenBank mRNA |
AK016629.1 |
- |
2, 3 |
373 |
|
AK016811.1 |
- |
3, 4 |
681 |
|
AK042750.1 |
- |
2, 3 |
555 |
|
AK077778.1 |
- |
2, 3 |
439 |
|
AK132428.1 |
- |
2, 3 |
522 |
|
BC053751.1 |
- |
2, 3 |
523 |
6335896 |
View Full Product Details