Gene Transcripts

Gene Symbol
Hnrnpll
Entrez Gene ID
72692
Gene Name
heterogeneous nuclear ribonucleoprotein L-like
Gene Aliases
2510028H02Rik, 2810036L13Rik, AI256697, AI852082, Hnrpll
Chromosome Location
Chr. 17: 80029487 - 80062513 on Build GRCm38
UniGene ID
Mm.64579
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144802.4 NP_659051.3 3 900
GenBank mRNA AK012866.1 - 3 616
AK034860.1 - 3 845
AK048900.1 - 3 689
AK146879.1 - 3 862
AK160943.1 - 2 708
AK171091.1 - 3 767
BC012849.1 - 3 846 3587716

View Full Product Details