Gene Transcripts

Gene Symbol
Prkag2
Entrez Gene ID
108099
Gene Name
protein kinase, AMP-activated, gamma 2 non-catalytic subunit
Gene Aliases
2410051C13Rik, AAKG, AAKG2, AI854673, H91620p, WPWS
Chromosome Location
Chr. 5: 24862735 - 25101906 on Build GRCm38
UniGene ID
Mm.33649
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170555.1 NP_001164026.1 7 620
NM_001170556.1 NP_001164027.1 7 617
NM_145401.2 NP_663376.2 11 1699
GenBank mRNA AK032238.1 - 9 1211
AK034765.1 - 5 285
AK154296.1 - 11 1700
AK165834.1 - 7 559
AY348864.1 - 7 550
BC015283.1 - 11 1487 4238045
BQ572978.1 - 7 550
BU707674.1 - 7 520

View Full Product Details