Gene Transcripts

Gene Symbol
Purg
Entrez Gene ID
75029
Gene Name
purine-rich element binding protein G
Gene Aliases
4930486B15Rik
Chromosome Location
Chr. 8: 33385614 - 33417472 on Build GRCm38
UniGene ID
Mm.159402
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098233.1 NP_001091703.1 1 546
NM_152821.2 NP_690034.1 1 546
GenBank mRNA AF479672.1 - 1 546
AF479673.1 - 1 546
AK015630.1 - 2 559
BC116406.2 - 1 546 40102393
CD772246.1 - 1 111

View Full Product Details