Gene Transcripts

Gene Symbol
Mfn1
Entrez Gene ID
67414
Gene Name
mitofusin 1
Gene Aliases
2310002F04Rik, 6330416C07Rik, D3Ertd265e, HR2, mKIAA4032
Chromosome Location
Chr. 3: 32529464 - 32579225 on Build GRCm38
UniGene ID
Mm.290414
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024200.4 NP_077162.2 4 596
GenBank mRNA AK018181.1 - 4 593
AK046313.1 - 4 549
AK141611.1 - 4 597
AK220169.1 - 4 584
AY174062.1 - 3 725
BC056641.1 - 4 555 5097345

View Full Product Details