Gene Transcripts

Gene Symbol
Rnpc3
Entrez Gene ID
67225
Gene Name
RNA-binding region (RNP1, RRM) containing 3
Gene Aliases
2810441O16Rik, AI447568, C030014B17Rik
Chromosome Location
Chr. 3: 113605067 - 113630149 on Build GRCm38
UniGene ID
Mm.316928
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038696.1 NP_001033785.1 7 733
NM_001287015.1 NP_001273944.1 7 733
NM_026043.3 NP_080319.2 7 733
GenBank mRNA AK013288.1 - 7 677
AK134241.1 - 7 698
AK161063.1 - 7 716
AK161974.1 - 7 686
AK163729.1 - 7 686
AK173272.1 - 7 484
BC016603.1 - 4 249 3587176
BC043695.1 - 7 702 5250774

View Full Product Details