Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026175.5 | NP_080451.4 | 12 | 2010 | |
GenBank mRNA | AK004763.1 | - | 12 | 1993 | |
AK008075.1 | - | 1 | 37 | ||
AK028829.1 | - | 12 | 1962 | ||
AK029095.1 | - | 12 | 1980 | ||
AK030115.1 | - | 4 | 536 | ||
AK030223.1 | - | 12 | 1997 | ||
AK134178.1 | - | 12 | 1988 | ||
AK160061.1 | - | 12 | 1993 | ||
BC010727.1 | - | 3 | 465 | ||
BC029753.1 | - | 12 | 1974 | 2651773 |