Gene Transcripts
- Gene Symbol
- Ccdc50
- Entrez Gene ID
- 67501
- Gene Name
- coiled-coil domain containing 50
- Gene Aliases
- C3orf6
- Chromosome Location
- Chr. 16: 27388765 - 27452218 on Build GRCm38
- UniGene ID
- Mm.258985
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001025615.3 |
NP_001020786.1
|
4, 5 |
465 |
|
NM_001289436.1 |
NP_001276365.1
|
5, 6 |
511 |
|
NM_026202.4 |
NP_080478.2
|
4, 5 |
465 |
|
GenBank mRNA |
AJ534985.1 |
- |
4, 5 |
332 |
|
AK012190.1 |
- |
4, 5 |
359 |
|
AK016827.1 |
- |
4, 5 |
440 |
|
AK077557.1 |
- |
4, 5 |
403 |
|
AK147820.1 |
- |
4, 5 |
416 |
|
AK151672.1 |
- |
4, 5 |
427 |
|
AK162434.1 |
- |
4, 5 |
442 |
|
AK165214.1 |
- |
4, 5 |
370 |
|
BC144895.1 |
- |
4, 5 |
444 |
9053443 |
BC144898.1 |
- |
5, 6 |
491 |
|
BC145933.1 |
- |
4, 5 |
444 |
40131236 |
BC145935.1 |
- |
4, 5 |
444 |
40131238 |
BY716501.1 |
- |
4, 5 |
440 |
|
CJ045424.1 |
- |
4, 5 |
469 |
|
View Full Product Details