Gene Transcripts

Gene Symbol
Patl2
Entrez Gene ID
67578
Gene Name
protein associated with topoisomerase II homolog 2 (yeast)
Gene Aliases
4930424G05Rik, Pat1a
Chromosome Location
Chr. 2: 122120103 - 122186189 on Build GRCm38
UniGene ID
Mm.150462
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026251.2 NP_080527.2 18 2191
GenBank mRNA AK010811.1 - 11 1259
AK015195.1 - 18 2195

View Full Product Details