Gene Transcripts

Gene Symbol
Rgs10
Entrez Gene ID
67865
Gene Name
regulator of G-protein signalling 10
Gene Aliases
2310010N19Rik
Chromosome Location
Chr. 7: 128373621 - 128419061 on Build GRCm38
UniGene ID
Mm.18635
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026418.2 NP_080694.1 4 353
GenBank mRNA AK009283.1 - 4 354
AK009686.1 - 4 355
AK017314.1 - 4 354
AK021202.1 - 4 354
AK088009.1 - 4 352
BC109178.1 - 4 338 40049527
BC109179.1 - 4 345

View Full Product Details