Gene Transcripts

Gene Symbol
Snx24
Entrez Gene ID
69226
Gene Name
sorting nexing 24
Gene Aliases
2810011K15Rik, 5730433I16Rik, AI451341
Chromosome Location
Chr. 18: 53244514 - 53390837 on Build GRCm38
UniGene ID
Mm.32414
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029394.3 NP_083670.1 7 860
GenBank mRNA AK017610.1 - 7 754
BC051168.1 - 7 863 6306904

View Full Product Details