Gene Transcripts

Gene Symbol
Vps26b
Entrez Gene ID
69091
Gene Name
vacuolar protein sorting 26 homolog B (yeast)
Gene Aliases
1810012I05Rik, 2310075A12Rik, AI848392
Chromosome Location
Chr. 9: 27004502 - 27030094 on Build GRCm38
UniGene ID
Mm.218717
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178027.4 NP_821170.1 3 988
GenBank mRNA AK010160.1 - 3 922
AK031589.1 - 3 991
AK150929.1 - 3 909
AK155578.1 - 3 949
AK165539.1 - 3 927
AK171154.1 - 3 947
AK215961.1 - 1 33
BC029758.1 - 1 81
BC049180.1 - 3 951 6314347
BC052721.1 - 3 912 6809853
BC062972.1 - 3 912 6809878

View Full Product Details