Gene Transcripts

Gene Symbol
Fam83g
Entrez Gene ID
69640
Gene Name
family with sequence similarity 83, member G
Gene Aliases
2310040C09Rik, AI506990, wly
Chromosome Location
Chr. 11: 61684082 - 61709950 on Build GRCm38
UniGene ID
Mm.458559
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178618.3 NP_848733.2 4 1343
GenBank mRNA AK028566.1 - 4 1290
BC085279.1 - 4 1304 30438273

View Full Product Details