Gene Transcripts

Gene Symbol
Tspan15
Entrez Gene ID
70423
Gene Name
tetraspanin 15
Gene Aliases
1110036D12Rik, 2700063A19Rik, AW048364, Tm4sf15
Chromosome Location
Chr. 10: 62185396 - 62231277 on Build GRCm38
UniGene ID
Mm.416781
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_197996.2 NP_932113.2 4 502
GenBank mRNA AK012471.1 - 4 501
AK172169.1 - 4 512
BC003872.1 - 4 451 3492667
CN455615.1 - 4 391
CX206586.1 - 4 502

View Full Product Details