Gene Transcripts

Gene Symbol
March7
Entrez Gene ID
57438
Gene Name
membrane-associated ring finger (C3HC4) 7
Gene Aliases
Axo, Axot, Gtrgeo17
Chromosome Location
Chr. 2: 60209892 - 60249211 on Build GRCm38
UniGene ID
Mm.260635
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020575.2 NP_065600.1 5 1202
GenBank mRNA AF155739.1 - 5 1202
AK034109.1 - 1192
AK143177.1 - 5 1224
AK166666.1 - 5 1209
AK167246.1 - 5 1184
AK168555.1 - 5 1205
BC025029.1 - 5 1166 4935124

View Full Product Details