Gene Transcripts

Gene Symbol
Srsf4
Entrez Gene ID
57317
Gene Name
serine/arginine-rich splicing factor 4
Gene Aliases
5730499P16Rik, AW550192, MNCb-2616, SRp75, Sfrs4
Chromosome Location
Chr. 4: 131873611 - 131901725 on Build GRCm38
UniGene ID
Mm.2478
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020587.2 NP_065612.2 6 2103
GenBank mRNA AB041587.1 - 3 1541
AK075769.1 - 6 2068
AK077648.1 - 6 2089
AK156989.1 - 6 2038
BC019437.1 - 6 2056 4018178
BC026944.1 - 6 2962 4016087
BC040812.1 - 6 2962

View Full Product Details