Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021547.3 | NP_067522.1 | 14 | 1405 | |
GenBank mRNA | AK041538.1 | - | 14 | 1361 | |
AK150166.1 | - | 14 | 1452 | ||
AK150254.1 | - | 14 | 1452 | ||
AK155124.1 | - | 14 | 1344 | ||
AK160480.1 | - | 13 | 2299 | ||
BC003313.1 | - | 14 | 1322 | 3155179 | |
BC058727.1 | - | 15 | 1492 | ||
X82457.1 | - | 14 | 1389 |