Gene Transcripts

Gene Symbol
Sav1
Entrez Gene ID
64010
Gene Name
salvador homolog 1 (Drosophila)
Gene Aliases
1700040G09Rik, Salv, Sav, WW45, Wwp3, Wwp4
Chromosome Location
Chr. 12: 69965012 - 69987002 on Build GRCm38
UniGene ID
Mm.390480
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022028.2 NP_071311.1 5 1721
GenBank mRNA AJ292968.1 - 5 1724
AK034983.1 - 3 2594

View Full Product Details